Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Blue Dextran
Synonym(s):
Dextran blue from
Leuconostoc mesenteroides
CAS Number:
87915-38-6
Products
Product Category
carbohydrates (4)
Brand
Sigma-Aldrich (6)
Biological Source
Physical Form
Product Line
Quality Segment
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
D5751
Blue Dextran,
Void volume marker for gel filtration columns.
Expand
View Pricing
D4772
Blue Dextran,
BioReagent
Expand
View Pricing
72762
Blue Dextran,
average mol wt 500,000
Expand
View Pricing
03714
Blue Dextran,
average mol wt 20,000
Expand
View Pricing
90008
Blue Dextran,
average mol wt 5,000
Expand
View Pricing
90475
Blue Dextran,
average mol wt 10,000
Expand
View Pricing