Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bismuth(III) nitrate pentahydrate
Linear Formula:
Bi(NO
3
)
3
· 5H
2
O
CAS Number:
10035-06-0
Molecular Weight:
485.07
EC Number:
233-791-8
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
ACS reagent (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
383074
Bismuth(III) nitrate pentahydrate,
ACS reagent, ≥98.0%
Expand
View Pricing
248592
Bismuth(III) nitrate pentahydrate,
reagent grade, 98%
Expand
View Pricing
467839
Bismuth(III) nitrate pentahydrate,
≥99.99% trace metals basis
Expand
View Pricing
254150
Bismuth(III) nitrate pentahydrate,
99.999% trace metals basis
Expand
View Pricing