Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bis(cyclopentadienyl)zirconium(IV) dichloride
Synonym(s):
Di(cyclopentadienyl)zirconium(IV) dichloride, Zirconocene dichloride
Empirical Formula (Hill Notation):
C
10
H
10
Cl
2
Zr
CAS Number:
1291-32-3
Molecular Weight:
292.32
EC Number:
215-066-8
Products
Brand
Sigma-Aldrich (1)
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Reaction Type
Reagent Type
Core
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
196215
Bis(cyclopentadienyl)zirconium(IV) dichloride,
≥98%
Expand
View Pricing