Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bioactive glass powder
Linear Formula:
CaO:SiO
2
:P
2
O
5
Products
Brand
Sigma-Aldrich (5)
Physical Form
Purity
Quality Segment
Particle Size (µm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
915084
Bioactive glass powder,
45S, ≥98%, 0.2-500 μm particle size
Expand
View Pricing
916323
Bioactive glass powder,
58S, ≥98%, ≤10 μm particle size
Expand
View Pricing
916072
Bioactive glass powder,
85S, ≥98%, 0.2-20 μm particle size
Expand
View Pricing
915076
Bioactive glass powder,
63S, ≥98%, 0.2-20 μm particle size
Expand
View Pricing
915335
Bioactive glass powder,
77S, ≥98%, 0.2-20 μm particle size
Expand
View Pricing