Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size
Products
Product Category
filter discs (19)
Brand
Millipore (19)
Manufacturer
Diameter (mm)
Hydrophobicity
Color
Membrane Material
Pore size (µm)
Sterility
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
AP2504700
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 47 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2004700
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 47 mm, hydrophilic, thickness 380 μm
Expand
View Pricing
AP2002500
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 25 mm, hydrophilic, thickness 380 μm
Expand
View Pricing
AP2001300
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 13 mm, hydrophilic, thickness 380 μm
Expand
View Pricing
AP2007500
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 75 mm, hydrophilic, thickness 380 μm
Expand
View Pricing
AP2502500
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 25 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2012450
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 124 mm, hydrophilic, thickness 380 μm
Expand
View Pricing
AP2009000
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 90 mm, hydrophilic, thickness 380 μm
Expand
View Pricing
AP2004200
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 42 mm, hydrophilic, thickness 380 μm
Expand
View Pricing
AP2512450
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 124 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2507500
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 75 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2501000
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 10 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2501300
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 13 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2514250
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 142 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2014250
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 142 mm, hydrophilic, thickness 380 μm
Expand
View Pricing
AP2502200
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 22 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2504200
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 42 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2509000
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 90 mm, hydrophilic, thickness 1.2 mm
Expand
View Pricing
AP2005500
Binder Resin Glass Fiber Membrane Filter, 2.0 μm Pore Size,
Millipore, filter diam. 55 mm, hydrophilic, thickness 380 μm
Expand
View Pricing