Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Glucuronidase from
Helix pomatia
Synonym(s):
β-D-Glucuronide glucuronosohydrolase
CAS Number:
9001-45-0
Products
Product Category
enzymes (3)
Brand
Sigma-Aldrich (7)
Biological Source
Physical Form
Quality Segment
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
G7017
β-Glucuronidase from
Helix pomatia
,
Type HP-2, aqueous solution, ≥100,000 units/mL
Expand
View Pricing
G0751
β-Glucuronidase from
Helix pomatia
,
Type H-1, partially purified powder, ≥300,000 units/g solid
Expand
View Pricing
G0876
β-Glucuronidase from
Helix pomatia
,
Type H-2, aqueous solution, ≥85,000 units/mL
Expand
View Pricing
G1512
β-Glucuronidase from
Helix pomatia
,
Type H-5, lyophilized powder, ≥400,000 units/g solid
Expand
View Pricing
G7770
β-Glucuronidase from
Helix pomatia
,
Type HP-2S, aqueous solution, ≥90,000 units/mL
Expand
View Pricing
G8885
β-Glucuronidase from
Helix pomatia
,
Type H-3, aqueous solution, ≥90,000 units/mL
Expand
View Pricing
G0762
β-Glucuronidase from
Helix pomatia
,
Type H-3AF, aqueous solution, ≥60,000 units/mL
Expand
View Pricing