Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benzo[
k
]fluoranthene
Empirical Formula (Hill Notation):
C
20
H
12
CAS Number:
207-08-9
Molecular Weight:
252.31
EC Number:
205-916-6
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
certified reference material (2)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
392251
Benzo[
k
]fluoranthene,
for fluorescence, ≥99%
Expand
View Pricing
03323
Benzo[
k
]fluoranthene,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing
BCR048R
Benzo[
k
]fluoranthene,
BCR
®
, certified reference material
Expand
View Pricing