Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benchmark Zirconium Bulk Beads
Synonym(s):
grinding medium, grinding beads, beads for homogenization
Products
Product Category
homogenizers (5)
Manufacturer
Diameter (mm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
BMSD113201TP
Benchmark Zirconium Bulk Beads,
bead diam. 0.1 mm, Molecular Biology (TriplePure)
Expand
View Pricing
BMSD113205TP
Benchmark Zirconium Bulk Beads,
bead diam. 0.5 mm, Molecular Biology (TriplePure)
Expand
View Pricing
BMSD113230TP
Benchmark Zirconium Bulk Beads,
bead diam. 3 mm, Molecular Biology (TriplePure)
Expand
View Pricing
BMSD113210TP
Benchmark Zirconium Bulk Beads,
bead diam. 1 mm, Molecular Biology (TriplePure)
Expand
View Pricing
BMSD113215TP
Benchmark Zirconium Bulk Beads,
bead diam. 1.5 mm, Molecular Biology (TriplePure)
Expand
View Pricing