Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
β-Asarone
Synonym(s):
cis
-1-Propenyl-2,4,5-trimethoxybenzene,
cis
-Asarone,
cis
-Isoasarone
Linear Formula:
(CH
3
O)
3
C
6
H
2
CH=CHCH
3
CAS Number:
5273-86-9
Molecular Weight:
208.25
EC Number:
226-096-6
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
analytical standard (1)
primary reference standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
PHL89568
β-Asarone,
phyproof
®
Reference Substance
Expand
View Pricing
02890590
β-Asarone,
primary reference standard
Expand
View Pricing