
Bacteriochlorophyll from Rhodopseudomonas sphaeroides

Synonym(s):
Bacteriochlorophyll a
Linear Formula:
C55H74N4O6Mg
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing