Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture
Products
Product Category
bottles (14)
Manufacturer
Capacity (mL)
Bottle material
Vial Bottle Material
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
DWKWGF600P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Distilled water, low-density polyethylene bottle, capacity 250 mL
Expand
View Pricing
DWKWGF541P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Ethanol, low-density polyethylene bottle, capacity 500 mL
Expand
View Pricing
DWKWGF531P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Acetone, low-density polyethylene bottle, capacity 250 mL
Expand
View Pricing
DWKWGF601P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Distilled water, low-density polyethylene bottle, capacity 500 mL
Expand
View Pricing
DWKWGF536P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Water, capacity 500 mL, low-density polyethylene bottle
Expand
View Pricing
DWKWGF539P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Isopropanol, low-density polyethylene bottle, capacity 500 mL
Expand
View Pricing
DWKWGF537P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Acetone, capacity 500 mL, low-density polyethylene bottle
Expand
View Pricing
DWKWGF533P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Isopropanol, low-density polyethylene bottle, capacity 250 mL
Expand
View Pricing
DWKWGF535P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Ethanol, low-density polyethylene bottle, capacity 250 mL
Expand
View Pricing
DWKWGF530P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Water, low-density polyethylene bottle, capacity 250 mL
Expand
View Pricing
DWKWGF532P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Methanol, low-density polyethylene bottle, capacity 250 mL
Expand
View Pricing
DWKWGF538P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, Methanol, low-density polyethylene bottle, capacity 500 mL
Expand
View Pricing
DWKWGF534P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, I.M.S, low-density polyethylene bottle, capacity 250 mL
Expand
View Pricing
DWKWGF540P
Azlon Pre-Labelled Narrow Neck Wash Bottles with jet-tip aperture,
label, I.M.S, low-density polyethylene bottle, capacity 500 mL
Expand
View Pricing