Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azlon Multi-Lingual Unvented Wash Bottles
Products
Product Category
bottles (20)
Manufacturer
Capacity (mL)
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
DWKWGW601PMLA
Azlon Multi-Lingual Unvented Wash Bottles,
label, Distilled water (in EN, FR, ES), capacity 500 mL
Expand
View Pricing
DWKWGW602PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Deionised water (in EN, FR, DE, ES), capacity 250 mL
Expand
View Pricing
DWKWGW701PP
Azlon Multi-Lingual Unvented Wash Bottles,
label, Dichloromethane label (in EN, FR, DE, ES), capacity 500 mL
Expand
View Pricing
DWKWGW539PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Isopropanol (in EN, FR, DE, ES, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW541PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Ethanol (in English, French, German, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW536PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Water (in English, French, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW601PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Distilled water (in EN, FR, DE, ES), capacity 500 mL
Expand
View Pricing
DWKWGW700PP
Azlon Multi-Lingual Unvented Wash Bottles,
capacity 500 mL
Expand
View Pricing
DWKWGW538PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Methanol (in EN, FR, DE, ES), capacity 500 mL
Expand
View Pricing
DWKWGW537PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Acetone (in English, French, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW600PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Distilled water (in EN, FR, DE, ES), capacity 250 mL
Expand
View Pricing
DWKWGW603PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Deionised (in EN, FR, DE, ES), capacity 500 mL
Expand
View Pricing
DWKWGW533PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Isopropanol (in English, French, Spanish), capacity 250 mL
Expand
View Pricing
DWKWGW540PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, I.M.S (in EN, FR, DE, ES), capacity 500 mL
Expand
View Pricing
DWKWGW703PP
Azlon Multi-Lingual Unvented Wash Bottles,
label, Methyl Ethyl Keton (in EN, FR, DE, ES), capacity 500 mL
Expand
View Pricing
DWKWGW530PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Water (in English, French, Spanish), capacity 250 mL
Expand
View Pricing
DWKWGW531PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Acetone (in English, French, Spanish), capacity 250 mL
Expand
View Pricing
DWKWGW532PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Methanol (in English, French, Spanish), capacity 250 mL
Expand
View Pricing
DWKWGW534PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, I.M.S (in English, French, Spanish), capacity 250 mL
Expand
View Pricing
DWKWGW535PML
Azlon Multi-Lingual Unvented Wash Bottles,
label, Ethanol (in English, French, Spanish), capacity 250 mL
Expand
View Pricing