Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting
Products
Product Category
bottles (30)
Manufacturer
Capacity (mL)
Filter Products
Showing 1-20 of 30
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
DWKWGW541VTML
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Ethanol (in English, French, German, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW537VTML
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Acetone (in English, French, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW537VTMLME
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Acetone (in English, French, Arabic), capacity 500 mL
Expand
View Pricing
DWKWGW541VTMLS
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Ethanol (in Norwegian, Danish, Swedish, English), capacity 500 mL
Expand
View Pricing
DWKWGW601PMLME
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Distilled water (in English, French, Arabic), capacity 500 mL
Expand
View Pricing
DWKWGW539VTMLA
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Isopropanol (in English, French, Spanish, OSHA), capacity 500 mL
Expand
View Pricing
DWKWGW601PMLS
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Distilled water (in Norwegian, Danish, Swedish, English), capacity 500 mL
Expand
View Pricing
DWKWGW537VTMLA
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Acetone (in English, French, Spanish, OSHA), capacity 500 mL
Expand
View Pricing
DWKWGW603PMLS
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Deionised water (in Norwegian, Danish, Swedish, English), capacity 500 mL
Expand
View Pricing
DWKWGW541VTMLA
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Ethanol (in English, French, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW532VTML
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Methanol (in English, French, Spanish), capacity 250 mL
Expand
View Pricing
DWKWGW531VTML
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Acetone (in English, French, Spanish), capacity 250 mL
Expand
View Pricing
DWKWGW538VTML
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Methanol (in English, French, German, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW540VTML
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, I.M.S (in English, French, German, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW537VTMLR
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Acetone (in Russian, Polish, German, English), capacity 500 mL
Expand
View Pricing
DWKWGW539VTML
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Isopropanol (in English, French, German, Spanish), capacity 500 mL
Expand
View Pricing
DWKWGW539VTMLS
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Isopropanol (in Norwegian, Danish, Swedish, English), capacity 500 mL
Expand
View Pricing
DWKWGW538VTMLA
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Methanol (in English, French, Spanish, OSHA), capacity 500 mL
Expand
View Pricing
DWKWGW538VTMLME
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Methanol (English, French, Arabic, Wide Neck), capacity 500 mL
Expand
View Pricing
DWKWGW538VTMLR
Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting,
label, Methanol (in Russian, Polish, German, English), capacity 500 mL
Expand
View Pricing
Page 1 of 2
Page 1 of 2