Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ART
®
SoftFit L Reload System
Products
Product Category
pipette tips (39)
Manufacturer
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-20 of 39
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z743892
ART
®
SoftFit L Reload System,
Pure 1000L, size 1000 μL, hinged rack, sterile
Expand
View Pricing
Z676861
ART
®
SoftFit L Reload System,
maximum volume 20 μL, Tip design ART 20L, Hinged rack, low retention: no
Expand
View Pricing
Z676896
ART
®
SoftFit L Reload System,
maximum volume 200 μL, Tip design ART 200L, Reload insert, low retention: no
Expand
View Pricing
Z676926
ART
®
SoftFit L Reload System,
maximum volume 200 μL, Tip design ART 200L, Hinged rack, low retention: no
Expand
View Pricing
Z677000
ART
®
SoftFit L Reload System,
maximum volume 1000 μL, Tip design ART 1000L, Hinged rack, low retention: no
Expand
View Pricing
Z676845
ART
®
SoftFit L Reload System,
maximum volume 20 μL, Tip design ART 20L, Reload insert, low retention: no
Expand
View Pricing
Z676969
ART
®
SoftFit L Reload System,
maximum volume 300 μL, Tip design ART 300L, Hinged rack, low retention: no
Expand
View Pricing
Z743907
ART
®
SoftFit L Reload System,
tip design ART 1200L, size 1200 μL, racked, sterile
Expand
View Pricing
Z676942
ART
®
SoftFit L Reload System,
maximum volume 300 μL, Tip design ART 300L, Reload insert, low retention: no
Expand
View Pricing
Z743906
ART
®
SoftFit L Reload System,
tip design ART 1000L, size 1000 μL, racked, sterile
Expand
View Pricing
Z676985
ART
®
SoftFit L Reload System,
maximum volume 1000 μL, Tip design ART 1000L, Reload insert, low retention: no
Expand
View Pricing
Z743913
ART
®
SoftFit L Reload System,
HLT 1000L, size 1000 μL, racked, non-sterile
Expand
View Pricing
Z677043
ART
®
SoftFit L Reload System,
maximum volume 1200 μL, Tip design ART 1200L, Hinged rack, low retention: no
Expand
View Pricing
Z743885
ART
®
SoftFit L Reload System,
HLT 200L, size 200 μL, racked, non-sterile
Expand
View Pricing
Z743889
ART
®
SoftFit L Reload System,
HLT 20L, size 20 μL, reload insert, non-sterile
Expand
View Pricing
Z743894
ART
®
SoftFit L Reload System,
Pure 1000L, size 1000 μL, reload insert, sterile
Expand
View Pricing
Z743882
ART
®
SoftFit L Reload System,
HLT 1200L, size 1200 μL, hinged rack, non-sterile
Expand
View Pricing
Z743908
ART
®
SoftFit L Reload System,
tip design ART 200L, size 200 μL, racked, sterile
Expand
View Pricing
Z743881
ART
®
SoftFit L Reload System,
HLT 1000L, size 1000 μL, hinged rack, non-sterile
Expand
View Pricing
Z743886
ART
®
SoftFit L Reload System,
HLT 200L, size 200 μL, reload insert, non-sterile
Expand
View Pricing
Page 1 of 2
Page 1 of 2