Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ART
®
low retention barrier tips
Products
Product Category
pipette tips (14)
Manufacturer
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
Z676764
ART
®
low retention barrier tips,
Tip design ART 200, Hinged rack
Expand
View Pricing
Z676578
ART
®
low retention barrier tips,
Tip design ART 10 REACH, Hinged rack
Expand
View Pricing
Z676659
ART
®
low retention barrier tips,
Tip design ART 20P, Hinged rack
Expand
View Pricing
Z676543
ART
®
low retention barrier tips,
Tip design ART 10 REACH, Reload insert
Expand
View Pricing
Z676802
ART
®
low retention barrier tips,
Tip design ART 1000XL, Hinged rack
Expand
View Pricing
Z676527
ART
®
low retention barrier tips,
Tip design ART 10, Hinged rack
Expand
View Pricing
Z676748
ART
®
low retention barrier tips,
Tip design ART 200, Reload insert
Expand
View Pricing
Z676594
ART
®
low retention barrier tips,
Tip design ART 20E, Reload insert
Expand
View Pricing
Z676616
ART
®
low retention barrier tips,
Tip design ART 20E, Hinged rack
Expand
View Pricing
Z676780
ART
®
low retention barrier tips,
Tip design ART 1000XL, Reload insert
Expand
View Pricing
Z676632
ART
®
low retention barrier tips,
Tip design ART 20P, Reload insert
Expand
View Pricing
Z676713
ART
®
low retention barrier tips,
Tip design ART 100E, Hinged rack
Expand
View Pricing
Z676500
ART
®
low retention barrier tips,
Tip design ART 10, Reload insert
Expand
View Pricing
Z676691
ART
®
low retention barrier tips,
Tip design ART 100E, Reload insert
Expand
View Pricing