Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ART
®
Barrier Tip
Products
Product Category
pipette tips (37)
Manufacturer
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-20 of 37
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z743699
ART
®
Barrier Tip,
ART 1000E, size 1000 μL, individually wrapped, sterile
Expand
View Pricing
Z743695
ART
®
Barrier Tip,
ART 1000 REACH, size 1000 μL, hinged rack, sterile
Expand
View Pricing
Z676799
ART
®
Barrier Tip,
maximum volume 1000 μL, Tip design ART 1000XL, Hinged rack, low retention: no
Expand
View Pricing
Z743706
ART
®
Barrier Tip,
ART 1250 REACH (extended length tip), size 1250 μL, hinged rack, sterile
Expand
View Pricing
Z676837
ART
®
Barrier Tip,
Empty reload rack, size , large, for 1000ul tips
Expand
View Pricing
Z676756
ART
®
Barrier Tip,
maximum volume 200 μL, Tip design ART 200, Hinged rack, low retention: no
Expand
View Pricing
Z676721
ART
®
Barrier Tip,
maximum volume 200 μL, Tip design ART 200, Reload insert, low retention: no
Expand
View Pricing
Z676640
ART
®
Barrier Tip,
maximum volume 20 μL, Tip design ART 20P, Hinged rack, low retention: no
Expand
View Pricing
Z676551
ART
®
Barrier Tip,
maximum volume 10 μL, Tip design ART 10 REACH, Hinged rack, low retention: no
Expand
View Pricing
Z743713
ART
®
Barrier Tip,
ART 200, size 200 μL, individually wrapped, sterile
Expand
View Pricing
Z676772
ART
®
Barrier Tip,
maximum volume 1000 μL, Tip design ART 1000XL, Reload insert, low retention: no
Expand
View Pricing
Z743718
ART
®
Barrier Tip,
ART 20P, size 200 μL, racked, sterile
Expand
View Pricing
Z743705
ART
®
Barrier Tip,
ART 1250 extended length, size 1250 μL, racked, sterile
Expand
View Pricing
Z676586
ART
®
Barrier Tip,
maximum volume 10 μL, Tip design ART 20E, Reload insert, low retention: no
Expand
View Pricing
Z676705
ART
®
Barrier Tip,
maximum volume 100 μL, Tip design ART 100E, Hinged rack, low retention: no
Expand
View Pricing
Z676519
ART
®
Barrier Tip,
maximum volume 10 μL, Tip design ART 10, Hinged rack, low retention: no
Expand
View Pricing
Z676608
ART
®
Barrier Tip,
maximum volume 10 μL, Tip design ART 20E, Hinged rack, low retention: no
Expand
View Pricing
Z676624
ART
®
Barrier Tip,
maximum volume 20 μL, Tip design ART 20P, Reload insert, low retention: no
Expand
View Pricing
Z676675
ART
®
Barrier Tip,
maximum volume 100 μL, Tip design ART 100, Hinged rack, low retention: no
Expand
View Pricing
Z676497
ART
®
Barrier Tip,
maximum volume 10 μL, Tip design ART 10, Reload insert, low retention: no
Expand
View Pricing
Page 1 of 2
Page 1 of 2