
Antithrombin III from human plasma

Synonym(s):
Factor Xa inhibitor, Heparin cofactor
CAS Number:
EC Number:
Products
Brand
Biological Source
Gene Alias
Physical Form
Purity
Tissue
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing