
Anti-Kindlin-1 antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-C20orf42, Monoclonal Anti-DTGCU2, Monoclonal Anti-FERMT1 URP1, Monoclonal Anti-KIND1, Monoclonal Anti-Kindlin-1 antibody produced in mouse, Monoclonal Anti-UNC112A, Monoclonal Anti-fermitin family member 1
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing