Anti-FBLN5 antibody produced in rabbit
Synonym(s):
Anti-Dance antibody produced in rabbit, Anti-Developmental arteries and neural crest EGF-like protein antibody produced in rabbit, Anti-FIBL-5 antibody produced in rabbit, Anti-Fibulin-5 precursor antibody produced in rabbit, Anti-UP50 antibody produced in rabbit, Anti-Urine p50 protein antibody produced in rabbit, Dance, Developmental arteries and neural crest EGF-like protein, FIBL-5, Fibulin-5, Urine p50 protein
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing