
Anisodine hydrobromide

Synonym(s):
α-Hydroxyscopolamin hydrobromide, Daturamine hydrobromide
Empirical Formula (Hill Notation):
C17H21NO5 · HBr
CAS Number:
Molecular Weight:
400.26
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing