Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Animal feeding needle, non-sterile, reusable, type 304 SS
Products
Product Category
needles (23)
Diameter (mm)
Length (in)
Filter Products
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
CAD7910
Animal feeding needle, non-sterile, reusable, type 304 SS,
curved, size 20G, L × diam. 1.5 in. × 2.25 mm , ball
Expand
View Pricing
CAD7912
Animal feeding needle, non-sterile, reusable, type 304 SS,
curved, size 18G, L × diam. 1.5 in. × 2.25 mm , ball
Expand
View Pricing
CAD7901
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 22G, L × diam. 1 in. × 1.25 mm , ball
Expand
View Pricing
CAD7921
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 20G, L × diam. 1 in. × 2.25 mm , ball
Expand
View Pricing
CAD7920
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 22G, L × diam. 1.5 in. × 1.25 mm , ball
Expand
View Pricing
CAD7902
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 20G, L × diam. 1.5 in. × 2.25 mm , ball
Expand
View Pricing
CAD7922
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 18G, L × diam. 1 in. × 2.25 mm , ball
Expand
View Pricing
CAD7923
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 16G, L × diam. 2 in. × 3 mm , ball
Expand
View Pricing
CAD7900
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 24G, L × diam. 1 in. × 1.25 mm , ball
Expand
View Pricing
CAD7911
Animal feeding needle, non-sterile, reusable, type 304 SS,
curved, size 20G, L × diam. 3 in. × 2.25 mm , ball
Expand
View Pricing
CAD7917
Animal feeding needle, non-sterile, reusable, type 304 SS,
curved, size 14G, L × diam. 3 in. × 4 mm , ball
Expand
View Pricing
CAD7914
Animal feeding needle, non-sterile, reusable, type 304 SS,
curved, size 18G, L 3 in. × 2.25 mm , ball
Expand
View Pricing
CAD7905
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 18G, L × diam. 2 in. × 2.25 mm , ball
Expand
View Pricing
CAD7906
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 18G, L × diam. 3 in. × 2.25 mm , ball
Expand
View Pricing
CAD7946
Animal feeding needle, non-sterile, reusable, type 304 SS,
curved, size 16G, L × diam. 4 × 3 mm , ball
Expand
View Pricing
CAD7913
Animal feeding needle, non-sterile, reusable, type 304 SS,
curved, size 18G, L × diam. 2 in. × 2.25 mm , ball
Expand
View Pricing
CAD7904
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 18G, L × diam. 1.5 in. × 2.25 mm , ball
Expand
View Pricing
CAD7919
Animal feeding needle, non-sterile, reusable, type 304 SS,
curved, size 12G, L × diam. 3.5 in. × 6.25 mm , ball
Expand
View Pricing
CAD7903
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 20G, L × diam. 3 in. × 2.25 mm , ball
Expand
View Pricing
CAD7908
Animal feeding needle, non-sterile, reusable, type 304 SS,
straight, size 16G, L × diam. 3 in. × 3 mm , ball
Expand
View Pricing
Page 1 of 2
Page 1 of 2