
Amorolfine hydrochloride

Synonym(s):
(2R,6S)-rel-4-[3-[4-(1,1-Dimethylpropyl)phenyl]-2-methylpropyl]-2,6-dimethyl-morpholine hydrochloride, Ro 14-4767/002
Empirical Formula (Hill Notation):
C21H35ON · HCl
CAS Number:
Molecular Weight:
353.97
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing