Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum oxide Inorganic Sorbent
Synonym(s):
Alumina
Linear Formula:
Al
2
O
3
CAS Number:
1344-28-1
Molecular Weight:
101.96
EC Number:
215-691-6
Products
Product Category
resins (1)
Brand
Supelco (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Separation Technique
Quality Segment
Pore Size (Å)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
06300
Aluminum oxide Inorganic Sorbent,
activated, neutral, 100-285 mesh, Brockmann Activity I
Expand
View Pricing