Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum heating/cooling block
Products
Manufacturer
Package Size
Technique
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
Z740270
Aluminum heating/cooling block,
Holds 96 x 0.2 mL tubes in center plus 10 on size (4 for 1.5ml tubes, 6 for 0.5ml tubes)
Expand
View Pricing
Z740273
Aluminum heating/cooling block,
Holds 80 x 1.5 mL tubes
Expand
View Pricing
Z740272
Aluminum heating/cooling block,
Holds 48 x 1.5 mL tubes
Expand
View Pricing
Z743474
Aluminum heating/cooling block,
for 0.2 mL PCR tubes/plates, 96-wells
Expand
View Pricing
Z743497
Aluminum heating/cooling block,
mini, for 1.5-2.0 mL tubes, 30 holes
Expand
View Pricing
Z743486
Aluminum heating/cooling block,
for 2.0 mL cryogenic tubes, 32-wells
Expand
View Pricing
Z743487
Aluminum heating/cooling block,
for 384-well plates, 384-wells in centre, 10 on sides (4 x 1.5 mL & 6 x 0.5 mL)
Expand
View Pricing
Z743480
Aluminum heating/cooling block,
for 1.5 mL cryogenic tubes, 55-wells
Expand
View Pricing
Z740271
Aluminum heating/cooling block,
Holds 192 x 0.2 mL tubes in center plus 20 on size (8 for 1.5ml tubes, 12 for 0.5ml tubes)
Expand
View Pricing
Z743492
Aluminum heating/cooling block,
for 50 mL tubes, 4 holes
Expand
View Pricing
Z743483
Aluminum heating/cooling block,
for 15.0 mL tubes, 12 holes
Expand
View Pricing
Z743477
Aluminum heating/cooling block,
for 0.5 mL tubes, 91-wells
Expand
View Pricing
Z743493
Aluminum heating/cooling block,
for flat-bottom microplates
Expand
View Pricing
Z743475
Aluminum heating/cooling block,
for 0.2 mL, 0.5 mL & 1.5 mL tubes, 79-wells
Expand
View Pricing
Z743490
Aluminum heating/cooling block,
for 5 mL tubes, 32-wells
Expand
View Pricing
Z743481
Aluminum heating/cooling block,
for 1.5 mL tubes, 64 holes, step block (4 rows of 16 holes)
Expand
View Pricing