Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7
Products
Manufacturer
Physical Form
Diameter (mm)
ID (mm)
Length (mm)
OD (mm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF10649178
Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7,
foil, Al 97.4%/Si 1%/Mg 0.9%/Mn 0.7%, thickness 1.0 mm, size 150 × 150 mm, temper t6
Expand
View Pricing
GF25030369
Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7,
200mm tube, 6.35mm outside diameter, 1.63mm wall thickness, 3.09mm inside diameter, solution treated
Expand
View Pricing
GF69989023
Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7,
500mm tube, 6.35mm outside diameter, 1.63mm wall thickness, 3.09mm inside diameter, solution treated
Expand
View Pricing
GF39538255
Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7,
1000mm tube, 6.35mm outside diameter, 1.63mm wall thickness, 3.09mm inside diameter, solution treated
Expand
View Pricing
GF94766619
Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7,
foil, thickness 1.0 mm, Al97.4%/Si 1%/Mg 0.9%/Mn 0.7%, size 300 × 300 mm, temper t6
Expand
View Pricing
GF58407789
Aluminum alloy 6082, Al97.4Si1Mg0.9Mn0.7,
100mm tube, 6.35mm outside diameter, 1.63mm wall thickness, 3.09mm inside diameter, solution treated
Expand
View Pricing