Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminium alloy 5754
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
GF93247555
Aluminium alloy 5754,
rod, Al97%/Mg 3%, 6.35 mm diameter, length 200 mm, condition extruded
Expand
View Pricing
GF03304774
Aluminium alloy 5754,
rod, Al 97%/Mg 3%, 6.35 mm diameter, length 100 mm, condition extruded
Expand
View Pricing
GF18576401
Aluminium alloy 5754,
rod, Al 97%/Mg 3%, 6.35 mm diameter, length 1000 mm, condition extruded
Expand
View Pricing
GF75960782
Aluminium alloy 5754,
rod, Al97%/Mg 3%, length 500 mm, 6.35 mm diameter, condition extruded
Expand
View Pricing