
All-rac-alpha-tocopheryl acetate for peak identification

Synonym(s):
DL-α-Tocopherol acetate, Vitamin E acetate, all-rac-α-Tocopheryl acetate
Empirical Formula (Hill Notation):
C31H52O3
CAS Number:
Molecular Weight:
472.74
Products
Product Category
Biological Source
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing