Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
NMR-tube cleaner
Products
Brand
Sigma-Aldrich (8)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z282391
Aldrich
®
NMR-tube cleaner,
Model C, joint: ST/NS 24/40 Model C, for tubes, 5 mm
Expand
View Pricing
Z282383
Aldrich
®
NMR-tube cleaner,
Model A, joint: tooled end; Model A, for tubes, 5 mm
Expand
View Pricing
Z174793
Aldrich
®
NMR-tube cleaner,
Model B, joint: ST/NS 24/40 Model B, for tubes, 5 mm
Expand
View Pricing
Z421553
Aldrich
®
NMR-tube cleaner,
Model C, joint: ST/NS 29/32, for tubes, 5 mm
Expand
View Pricing
Z239224
Aldrich
®
NMR-tube cleaner,
Model D, joint: ST/NS 29/32, for tubes, 5 mm
Expand
View Pricing
Z127418
Aldrich
®
NMR-tube cleaner,
Model C, joint: ST/NS 24/40, for tubes, 10 mm
Expand
View Pricing
Z220078
Aldrich
®
NMR-tube cleaner,
Model E, joint: ST/NS 24/40, for tubes, 5 mm
Expand
View Pricing
Z239216
Aldrich
®
NMR-tube cleaner,
Model D, joint: ST/NS 24/40, for tubes, 5 mm
Expand
View Pricing