Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
low form dewar flask
Products
Product Category
flasks (10)
Brand
Sigma-Aldrich (9)
Manufacturer
Capacity (mL)
Package Size
Sterility
Style
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z107549
Aldrich
®
low form dewar flask,
capacity 600 mL, hemispherical, depth 6 1/2 cm
Expand
View Pricing
Z202657
Aldrich
®
low form dewar flask,
capacity 850 mL, cylindrical, depth 7 1/2 cm
Expand
View Pricing
Z107530
Aldrich
®
low form dewar flask,
capacity 275 mL, hemispherical, depth 5 1/2 cm
Expand
View Pricing
Z183881
Aldrich
®
low form dewar flask,
capacity 100 mL, hemispherical, depth 3 cm
Expand
View Pricing
Z202630
Aldrich
®
low form dewar flask,
capacity 150 mL, cylindrical, depth 3 1/2 cm
Expand
View Pricing
Z202649
Aldrich
®
low form dewar flask,
capacity 350 mL, cylindrical, depth 7 1/2 cm
Expand
View Pricing
Z107565
Aldrich
®
low form dewar flask,
capacity 550 mL, cylindrical, inside depth 9 cm
Expand
View Pricing
Z150398
Aldrich
®
low form dewar flask,
capacity 150 mL, hemispherical, depth 4 cm
Expand
View Pricing
Z202665
Aldrich
®
low form dewar flask,
capacity 2,500 mL, cylindrical, depth 13 1/2 cm
Expand
View Pricing
Z107557
Aldrich
®
low form dewar flask,
capacity 220 mL, cylindrical, depth 6 1/2 cm
Expand
View Pricing