Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aldrich
®
filtering flask with hose barb, heavy-wall
Products
Product Category
flasks (9)
Brand
Sigma-Aldrich (9)
Manufacturer
Capacity (mL)
Package Size
Sterility
Style
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z568309
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity 500 mL, neck joint: ST/NS 24/40
Expand
View Pricing
Z568317
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity 1,000 mL, neck joint: ST/NS 24/40
Expand
View Pricing
Z568341
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity 250 mL
Expand
View Pricing
Z568333
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity 4,000 mL, neck joint: ST/NS 24/40
Expand
View Pricing
Z568325
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity 2,000 mL, neck joint: ST/NS 24/40
Expand
View Pricing
Z568376
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity × H 1,000 mL × 255 mm, neck joint: ST/NS 29/32
Expand
View Pricing
Z568295
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity 250 mL, neck joint: ST/NS 24/40
Expand
View Pricing
Z568384
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity 2,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z568392
Aldrich
®
filtering flask with hose barb, heavy-wall,
capacity 4,000 mL, neck joint: ST/NS 29/32
Expand
View Pricing