Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
α-Glucosidase from
Saccharomyces cerevisiae
Synonym(s):
α-D-Glucosidase, α-D-Glucoside glucohydrolase, Maltase from yeast
CAS Number:
9001-42-7
EC Number:
232-604-7
Products
Product Category
glycosidases (1)
Brand
Sigma-Aldrich (2)
Sustainability
Feature
Gene Alias
Physical Form
Recombinant Host
Chemical Composition
Quality Segment
Relevant Disease
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G5003
α-Glucosidase from
Saccharomyces cerevisiae
,
Type I, lyophilized powder, ≥10 units/mg protein (using p-nitrophenyl α-D-glucoside as substrate.)
Expand
View Pricing
G0660
α-Glucosidase from
Saccharomyces cerevisiae
,
recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein
Expand
View Pricing