Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Adapter for Eppendorf
®
Rotor S-4xUniversal-Large
Synonym(s):
Adapter for Eppendorf
®
S-4x Universal Large Rotor
Products
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP5920757008
Adapter for Eppendorf
®
Rotor S-4xUniversal-Large,
5/15mL conical tubes and plates, rotor S-4xUniversal Large
Expand
View Pricing
EP5910763000
Adapter for Eppendorf
®
Rotor S-4xUniversal-Large,
0.5-2mL micro test tubes, rotor S-4xUniversal
Expand
View Pricing
EP5920755005
Adapter for Eppendorf
®
Rotor S-4xUniversal-Large,
50mL conical tubes, 175-250 mL bottles and plates, 175-250 mL bottles and plates, rotor S-4xUniversal Large
Expand
View Pricing
EP5920756001
Adapter for Eppendorf
®
Rotor S-4xUniversal-Large,
25/50mL conical tubes and plates, rotor S-4xUniversal Large
Expand
View Pricing