Skip to Content
Merck
Acridine Orange base Dye content 75 %

Acridine Orange base

Acridine Orange base Dye content 75 %
Synonym(s):
3,6-Bis(dimethylamino)acridine, Solvent Orange 15
Empirical Formula (Hill Notation):
C17H19N3
CAS Number:
Molecular Weight:
265.35

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing