Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ace receiving flask,
Products
Product Category
flasks (11)
Manufacturer
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z682829
Ace receiving flask,,
capacity 1000 mL, joint: ST/NS 35/20, glass
Expand
View Pricing
Z682861
Ace receiving flask,,
capacity 1000 mL, joint: ST/NS 35/20, plastic-coated glass
Expand
View Pricing
Z682802
Ace receiving flask,,
capacity 250 mL, joint: ST/NS 35/20, glass
Expand
View Pricing
Z682845
Ace receiving flask,,
capacity 3000 mL, joint: ST/NS 35/20, glass
Expand
View Pricing
Z682799
Ace receiving flask,,
capacity 100 mL, joint: ST/NS 35/20, glass
Expand
View Pricing
Z682810
Ace receiving flask,,
capacity 500 mL, joint: ST/NS 35/20, glass
Expand
View Pricing
Z682837
Ace receiving flask,,
capacity 2000 mL, joint: ST/NS 35/20, glass
Expand
View Pricing
Z682888
Ace receiving flask,,
capacity 2000 mL, joint: ST/NS 35/20, plastic-coated glass
Expand
View Pricing
Z682896
Ace receiving flask,,
capacity 3000 mL, joint: ST/NS 35/20, plastic-coated glass
Expand
View Pricing
Z682853
Ace receiving flask,,
capacity 500 mL, joint: ST/NS 35/20, plastic-coated glass
Expand
View Pricing
Z682780
Ace receiving flask,,
capacity 50 mL, joint: ST/NS 35/20, glass
Expand
View Pricing