Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
ABC Vial
™
, 2 mL, preassembled, 9 mm thread
Products
Product Category
vials (5)
Brand
Supelco (5)
ID (mm)
OD (mm)
Volume (ml)
Closure Type
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
27490-U
ABC Vial
™
, 2 mL, preassembled, 9 mm thread,
clear glass vial (with marking spot), PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
27487-U
ABC Vial
™
, 2 mL, preassembled, 9 mm thread,
clear glass vial, PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
27548-U
ABC Vial
™
, 2 mL, preassembled, 9 mm thread,
clear glass vial (with marking spot), PTFE/silicone septum, pkg of 1000 ea
Expand
View Pricing
27496
ABC Vial
™
, 2 mL, preassembled, 9 mm thread,
amber glass vial (with marking spot), PTFE/silicone septum, pkg of 100 ea
Expand
View Pricing
27498
ABC Vial
™
, 2 mL, preassembled, 9 mm thread,
amber glass vial (with marking spot), PTFE/silicone/PTFE septum, pkg of 100 ea
Expand
View Pricing