
9-Aminoacridine hydrochloride monohydrate

Empirical Formula (Hill Notation):
C13H10N2 · HCl · H2O
Products
Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing