Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Azacytidine
Synonym(s):
4-Amino-1-(β-D-ribofuranosyl)-1,3,5-triazin-2(1
H
)-one, Ladakamycin
Empirical Formula (Hill Notation):
C
8
H
12
N
4
O
5
CAS Number:
320-67-2
Molecular Weight:
244.20
EC Number:
206-280-2
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Sterilization Method
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
A2385
5-Azacytidine,
≥98% (HPLC), powder, DNA methyltransferase inhibitor
Expand
View Pricing
A1287
5-Azacytidine,
Hybri-Max
™
, γ-irradiated, lyophilized powder, BioXtra, suitable for hybridoma
Expand
View Pricing