Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
5-Aminolevulinic acid hydrochloride
Synonym(s):
δ-Aminolevulinic acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, 5-Aminolaevulinic acid hydrochloride, ALA
Linear Formula:
NH
2
CH
2
C(O)CH
2
CH
2
COOH · HCl
CAS Number:
5451-09-2
Molecular Weight:
167.59
EC Number:
226-679-5
Products
Product Category
amino acids (2)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
Purum (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A3785
5-Aminolevulinic acid hydrochloride,
≥98%, suitable for ligand binding assays
Expand
View Pricing
A7793
5-Aminolevulinic acid hydrochloride,
BioReagent, suitable for cell culture, powder, ≥98%
Expand
View Pricing
08339
5-Aminolevulinic acid hydrochloride,
≥97.0% (AT)
Expand
View Pricing