
4-(Methylthio)butanol

Synonym(s):
4-(Methylthio)-1-butanol
Linear Formula:
CH3S(CH2)4OH
CAS Number:
Molecular Weight:
120.21
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing