Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
4-Carvomenthenol
Synonym(s):
4-Carvomenthenol, 4-Terpinenol
Empirical Formula (Hill Notation):
C
10
H
18
O
CAS Number:
562-74-3
Molecular Weight:
154.25
EC Number:
209-235-5
Products
Product Category
aroma chemicals (2)
Brand
Sigma-Aldrich (2)
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Food Grade (2)
Halal (2)
Fragrance grade (1)
Kosher (1)
Natural (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
W224820
4-Carvomenthenol,
natural, ≥95%, FG
Expand
View Pricing
W224847
4-Carvomenthenol,
≥95%, FCC, FG
Expand
View Pricing