Skip to Content
Products
Cart
0
AF
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
3-Phosphoglyceric Phosphokinase from baker’s yeast (
S. cerevisiae
)
Synonym(s):
3-Phosphoglycerate kinase, ATP:3-Phospho-D-glycerate 1-phosphotransferase
CAS Number:
9001-83-6
EC Number:
232-636-1
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (1)
Biological Source
Physical Form
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
P7634
3-Phosphoglyceric Phosphokinase from baker’s yeast (
S. cerevisiae
),
ammonium sulfate suspension, ≥500 units/mg protein
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More