Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(3-Aminopropyl)triethoxysilane
Synonym(s):
3-Triethoxysilylpropylamine, APTES, APTS
Linear Formula:
H
2
N(CH
2
)
3
Si(OC
2
H
5
)
3
CAS Number:
919-30-2
Molecular Weight:
221.37
EC Number:
213-048-4
Products
Product Category
adsorbents (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
440140
(3-Aminopropyl)triethoxysilane,
99%
Expand
View Pricing
741442
(3-Aminopropyl)triethoxysilane,
≥98.0%
Expand
View Pricing
A3648
(3-Aminopropyl)triethoxysilane,
≥98%
Expand
View Pricing
706493
(3-Aminopropyl)triethoxysilane,
packaged for use in deposition systems, ≥98%
Expand
View Pricing