Skip to Content
Merck
3,3′-Dihexyloxacarbocyanine iodide 98%

3,3′-Dihexyloxacarbocyanine iodide

3,3′-Dihexyloxacarbocyanine iodide 98%
Synonym(s):
3-hexyl-2-[3-(3-hexyl-2(3H)benzoxazolylidene)-1-propenyl]benzoxazolium iodide
Empirical Formula (Hill Notation):
C29H37IN2O2
CAS Number:
Molecular Weight:
572.52

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing