Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
3,3′,5,5′-Tetrabromobisphenol A
Synonym(s):
4,4′-Isopropylidenebis(2,6-dibromophenol), TBBPA
Linear Formula:
(CH
3
)
2
C[C
6
H
2
(Br)
2
OH]
2
CAS Number:
79-94-7
Molecular Weight:
543.87
EC Number:
201-236-9
Products
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Monomer Type
Special Grade
certified reference material (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
330396
3,3′,5,5′-Tetrabromobisphenol A,
97%
Expand
View Pricing
90758
3,3′,5,5′-Tetrabromobisphenol A,
certified reference material,
Trace
CERT
®
, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing