Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Naphthaleneacetic acid
Synonym(s):
2-Naphthylacetic acid
Linear Formula:
C
10
H
7
CH
2
CO
2
H
CAS Number:
581-96-4
Molecular Weight:
186.21
EC Number:
209-475-0
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
N4002
2-Naphthaleneacetic acid,
99%
Expand
View Pricing