Skip to Content
Merck
2-Methoxycinnamaldehyde phyproof® Reference Substance

2-Methoxycinnamaldehyde

2-Methoxycinnamaldehyde phyproof® Reference Substance
Synonym(s):
o-Methoxycinnamaldehyde
Linear Formula:
CH3OC6H4CH=CHCHO
CAS Number:
Molecular Weight:
162.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing