Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(2-Hydroxypropyl)-γ-cyclodextrin
Synonym(s):
HGC
CAS Number:
128446-34-4
Products
Product Category
polymers (1)
polysaccharides (1)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
pH Value
Physical Form
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
779229
(2-Hydroxypropyl)-γ-cyclodextrin,
Wacker Chemie AG
Expand
View Pricing
390704
(2-Hydroxypropyl)-γ-cyclodextrin,
extent of labeling: 0.6 molar substitution
Expand
View Pricing
H125
(2-Hydroxypropyl)-γ-cyclodextrin,
solid
Expand
View Pricing