Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Hydroxyethyl methacrylate
Synonym(s):
1,2-Ethanediol mono(2-methylpropenoate), Glycol methacrylate, HEMA, 2-Hydroxyethyl methacrylate
Linear Formula:
CH
2
=C(CH
3
)COOCH
2
CH
2
OH
CAS Number:
868-77-9
Molecular Weight:
130.14
EC Number:
212-782-2
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
128635
2-Hydroxyethyl methacrylate,
contains ≤250 ppm monomethyl ether hydroquinone as inhibitor, 97%
Expand
View Pricing
477028
2-Hydroxyethyl methacrylate,
≥99%, contains ≤50 ppm monomethyl ether hydroquinone as inhibitor
Expand
View Pricing
8.00588
2-Hydroxyethyl methacrylate,
(stabilised with hydroquinone monomethyl ether) for synthesis
Expand
View Pricing
17348
2-Hydroxyethyl methacrylate,
embedding medium (for microscopy)
Expand
View Pricing