Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Chloro-4-fluorotoluene
Linear Formula:
CH
3
C
6
H
3
(Cl)F
CAS Number:
452-73-3
Molecular Weight:
144.57
EC Number:
207-209-8
Products
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Special Grade
certified reference material (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
80730
2-Chloro-4-fluorotoluene,
Trace
CERT
®
, certified reference material,
19
F-qNMR Standard, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing