Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
2-Bromoethylamine hydrobromide
Synonym(s):
2-Aminoethyl bromide hydrobromide
Linear Formula:
BrCH
2
CH
2
NH
2
· HBr
CAS Number:
2576-47-8
Molecular Weight:
204.89
EC Number:
219-924-2
Products
Product Category
building blocks (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Purum (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
B65705
2-Bromoethylamine hydrobromide,
99%
Expand
View Pricing
06670
2-Bromoethylamine hydrobromide,
purum, ≥97.0% (AT)
Expand
View Pricing